Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Catalog # APOSOR13646-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, Methyl 3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-carboxylate
:
:223.25 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:188614-01-9
Formula:C₁₀H₉NO₃S
MDL Number:MFCD00449104
Melting Pt:97…99 °C