1-Ethynyl-1-cyclohexene
Catalog # APOSOR13588-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: (1-Cyclohexenyl)acetylene, 1-Ethynylcyclohexene
:
:106.17 g/mol
Storage Temperature:Ambient
CAS Number:931-49-7
Formula:C₈H₁₀
MDL Number:MFCD00001568