Order Entry
Export
ContactUsLinkComponent
Methyl-3-hydroxy-4-iodobenzoate
Methyl-3-hydroxy-4-iodobenzoate
Catalog # APOSOR111325-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-3-hydroxy-4-iodobenzoate
Catalog # APOSOR111325-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 3-hydroxy-4-iodobenzoate
 :
 :278.05 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:157942-12-6
Formula:C₈H₇IO₃
MDL Number:MFCD06203960