2-Bromo-5-fluorobenzenesulphonyl chloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Bromo-5-fluorobenzene-1-sulphonyl chloride
CAS Number:771-67-5
:
Formula:C₆H₃BrClFO₂S
MDL Number:MFCD13185361
Molecular Weight:273.507 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: