7-Oxabicyclo[4.1.0]heptan-2-one 95%
Catalog # APOSOR939492-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:112.127 g/mol
Storage Temperature:Ambient
CAS Number:6705-49-3
Formula:C₆H₈O₂
MDL Number:MFCD00192371