N-Methyl-3-(triethoxysilyl)propylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: N-Methyl-3-(triethoxysilyl)propan-1-amine, [3-(Methylamino)prop-1-yl]triethoxysilane
CAS Number:6044-50-4
:
Formula:C₁₀H₂₅NO₃Si
MDL Number:MFCD24842713
Boiling Pt:120…121 °C (30 mm Hg)
Molecular Weight:235.396 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: