Order Entry
Export
ContactUsLinkComponent
Methylene blue, tablets for milk analysis
Methylene blue, tablets for milk analysis
Catalog # 330733R
Supplier:  VWR Chemicals
CAS Number:  
undefined
Methylene blue, tablets for milk analysis
Catalog # 330733R
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Aluminium tube
  • Pk:
    50 Tab.
  • Seq:
    0935
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance::
    .
  • Purple FBE or flat tablets:
    Passes test
  • Matter insoluble in water:
    Passes test
  • Weight uniformity:
    Passes test
  • Average tablet weight:
    437 --> 483 mg
  • Diameter:
    11.0 --> 11.5 mm
  • Hardness (kp):
    Passes test
  • Methylene blue (mg/tablet):
    14.2 --> 17.4

Specification Test Results

About this item

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
 :
 :319.86 g/mol
EINECS:200-515-2
 :GHS07
Storage Temperature:Ambient
CAS Number:61-73-4
 :MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008
Melting Pt:180
Density:1.757 g/cm³ (22 °C)
Merck Index:12,06137