3,4,5-Trimethoxybenzoyl chloride, Sigma-Aldrich®
Catalog # SIALT69809-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3,4,5-Trimethoxybenzene-1-carbonyl chloride
:
:230.65 g/mol
EINECS:224-851-4
UN:3261
:GHS05
CAS Number:4521-61-3
Formula:C₁₀H₁₁ClO₄
MDL Number:MFCD00000675
Boiling Pt:315
Melting Pt:90…91 °C
Density:∼5.8 g/cm³ (20 °C)
ADR:8 II
:M-Clarity™ quality level = MQ100