Arsenic limit test apparatus
NEUB23900031002920
- Description:Arsenic limit test apparatus, complete, without flange, NS 29/32, piston content (100 ml)
- Environmentally Preferable:
- Pk:1
This apparatus is used for the determination of the limit of arsenic.
Complete unit with four parts with springs, attachment and tension springs.
: Pharmeuropa 28.2; Ph.Eur 2.4.2 method A, EP 9.4