trans-4-(Trifluoromethyl)cinnamic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16642-92-5
:
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD00002696
Molecular Weight:216.16 g/mol
Melting Pt:230…234 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: