Nitrile gloves, Edge® 82-133
Supplier: Ansell
Certificates
About this item
These disposable gloves are manufactured with a thinner nitrile formulation that increases comfort and improves flexibility without compromising protection.
- Beaded cuffs for easy donning and removal
- Textured fingertips provide an improved grip for a variety of applications
- Packed 300 gloves per dispenser box to provide additional value and convenience (250 pieces/dispenser for size XL)
Typical applications include: Catering, compounding, mixing, blending, fastening, screwing & unscrewing parts, food processing, manufacturing, inspection and tooling precision maintenance, lab work, blending, compounding, filling, cleaning, and maintenance and equipment cleanup.
Certifications: EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 VIRUS; Cat. III
Food contact approved
Food contact approved