6-Bromohexanoyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:22809-37-6
:
Formula:BrH₂C(CH₂)₄COOCl
MDL Number:MFCD00000761
Boiling Pt:130
Density:1.38 g/cm³ (20 °C)
Molecular Weight:213.5 g/mol
Storage Temperature:Ambient
EINECS:245-236-7
UN:3265
ADR:8 II
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable: