L(+)-Leucinol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: L-Leucinol, (S)-(+)-2-Amino-4-methylpentan-1-ol, (S)-2-Amino-4-methylpentan-1-ol
CAS Number:7533-40-6
:
Formula:C₆H₁₅NO
MDL Number:MFCD00063676
Boiling Pt:198
Melting Pt:66…70 °C
Density:0.91 g/cm³
Molecular Weight:117.19 g/mol
Flash Pt:90 °C
EINECS:231-400-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4490 to 1.4530 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:+3° to +5° (20°C, 589 nm) (c=9, ethanol)