ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
  ROCH10789704001
 :  ROCHE
 :  
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
  ROCH10789704001
 :  ROCHE
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:
 : Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
 :
 :3082
 :GHS05
 :GHS07
 :GHS09
 :∼200 °C (1013 hPa)
 :251 °C (closed cup)
 :Refrigerator
 :9002-93-1
 :9036-19-5
 :MFCD00132505
 :6
 :1.06 g/cm³ (20 °C)
 :9 III
 :13,06793
 :M-Clarity™ quality level = MQ100