Order Entry
Export
ContactUsLinkComponent
 
ATP (Energy) Regeneration Solution
Catalog # ENZOBMLEW98100100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
ATP (Energy) Regeneration Solution
Catalog # ENZOBMLEW98100100
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Enzyme name:
    ATP (Energy) regeneration solution

Specifications

About this item

This solution contains MgCl₂, ATP, and ATP regenerating enzymes to recycle ATP hydrolysis products, (AMP, ADP) to ATP. An ideal energy source for conjugation/degradation assays (ubiquitin, proteasome, SUMO, etc.) particularly those that are preformed with crude lysates/impure reagents that may contain ATP-depleting activities.

Liquid. In 50 mM TRIS-Cl, pH 7,5.