Bovine fetuin (from Fetal serum)
: Merck
Fetuin extracted from foetal bovine serum. Fetuin is an α-globulin that inhibits trypsin activity and promotes cell attachment, growth and differentiation in many different culture systems.
- Lyophilized powder
- Impurities ≤0,3% free N-acetylneuraminic acid
- Source:
- Species:
- Environmentally Preferable: