Anti-PRKD1 Rabbit Polyclonal Antibody
Catalog # PRSI79-551
Supplier: ProSci Inc.
Specifications
- Antigen name:Protein Kinase D1
- Antigen symbol:PRKD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:PRKCM|Protein kinase D|nPKC-D1|PKD|Serine/threonine-protein kinase D1|PKD1|Protein kinase C mu type|PRKD1|nPKC-mu
- Concentration:1 mg/mL
- Immunogen:PKD/PKCmu (Ab-910) antibody was raised against a peptide sequence around aa. 908~912 (R-V-S-I-L) derived from Human PKD/PKCμ.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Converts transient diacylglycerol (DAG) signals into prolonged physiological effects, downstream of PKC. Involved in resistance to oxidative stress through activation of NF-κ-B.
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat