Anti-AUTS2 Goat Polyclonal Antibody
Catalog # PRSI46-970
Supplier: ProSci Inc.
Specifications
- Antigen name:Autism Susceptibility Candidate 2
- Antigen symbol:AUTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:AUTS2
- Concentration:500 ug/mL
- Immunogen:AUTS2 antibody was raised against a 16 amino acid synthetic peptide near the C-Terminus of AUTS2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
AUTS2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
AUTS2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AUTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human