Anti-ANAPC7 Rabbit Polyclonal Antibody
Catalog # PRSI29-605
Supplier: ProSci Inc.
Specifications
- Antigen name:Anaphase Promoting Complex Subunit 7
- Antigen symbol:ANAPC7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:APC7
- Immunogen:A synthetic peptide corresponding a region of human ANAPC7.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The anaphase-promoting complex (APC) consists of at least 8 protein subunits, including APC5, CDC27 (APC3), CDC16 (APC6), and CDC23 (APC8).The anaphase-promoting complex (APC) consists of at least 8 protein subunits, including APC5, CDC27 (APC3; MIM 116946), CDC16 (APC6; MIM 603461), and CDC23 (APC8; MIM 603462).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ANAPC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse