ContactUsLinkComponent
Gas sampling pump and tubes
Gas sampling pump and tubes
Catalog # 763-0292
Supplier:  GASTEC
undefined
Gas sampling pump and tubes
Catalog # 763-0292
Supplier:  GASTEC
Specifications
  • Description:
    Gas sampling pump for science education (50 cm3)
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Gas detector tube system accurately measures vapour concentration levels and slight changes in concentration level.
These detector tubes are used in experiments like respiration, photosynthesis and combustion to determine concentration level.
Ordering information: Includes a sampling pump, detector tubes and tips for detector tubes. More detector tubes are available separately.
Gas Tubes
Product Information