Oleic acid Ph. Eur., Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (9Z)-9-Octadecenoic acid
CAS Number:112-80-1
:
Formula:C₁₈H₃₄O₂
MDL Number:MFCD00064242
Boiling Pt:286
Molecular Weight:282.47 g/mol
Melting Pt:16.7
Flash Pt:180
Merck Index:13,06898
:M-Clarity™ quality level = MQ200
Specifications
- Environmentally Preferable: