ContactUsLinkComponent
Hydrogen flashback arrestor
Hydrogen flashback arrestor
  REST21334
 :  Restek
undefined
Hydrogen flashback arrestor
  REST21334
 :  Restek
  • Description:
    Hydrogen flashback arrestor, brass body, 6,35 mm female N fittings
  • Environmentally Preferable:
  • Pk:
    1
This hydrogen flashback arrestor is a special gas safety device, it cuts to stop the flame or reverse flow of gas back up into the equipment or supply line and it prevents the user and equipment from damage or explosions.
  • Valve cuts off gas supply in the event of a flashback
  • Can be reset and reused after flashback has occurred
  • Valve spring is made of 316 stainless steel
Maximum pressure: Hydrogen, 3,5 bar, temperature sensor: polyethylene
 : Meets safety requirements of BS6158, EN730, ISO 5175, AS 4603 and are UL Listed 23Y5.