Order Entry
Export
ContactUsLinkComponent
 
Anti-NEFL Mouse Monoclonal Antibody (Purified) [clone: NR-4]
 :  Biotium
 
Anti-NEFL Mouse Monoclonal Antibody (Purified) [clone: NR-4]
 :  Biotium

 

  • Catalog No:
  • BNUB0303-100
  • BNUB0303-500
  • Antigen symbol:
  • NEFL
  • NEFL
  • Antigen name:
  • Neurofilament
  • Neurofilament
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone NR-4
  • Clone NR-4
  • Reactivity:
  • Pig, Chicken, Human, Bovine, Rat
  • Pig, Chicken, Human, Bovine, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 521461
  • 521461
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide