Lymphocyte separation medium, reagent grade, LSM™, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Lymphocyte Separation Medium (LSM™) is formulated for isolation of mononuclear cells from defibrinated or heparinised whole human blood. One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets. It is a sterile-filtered solution of a sucrose polymer and diatrizoate salts.