1-Boc-3-oxopiperazine 97%
: tert-butyl 3-oxopiperazine-1-carboxylate, tert-butyl 3-oxo-1-piperazinecarboxylate, 1-Boc-3-Oxopiperazine, 4-N-BOC-2-oxo-piperazine, 4-Boc-2-Oxopiperazine
:76003-29-7
:
:C₉H₁₆N₂O₃
:MFCD02181069
:200.238 g/mol
:160…163 °C
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:159 to 164 °C
- GC:≥96.0 %