1-Bromo-2-(methoxymethyl)benzene 98%
: 1-Bromobenzyl methyl ether
:52711-30-5
:
:C₈H₉BrO
:MFCD03790887
:201.0605 g/mol
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5470 to 1.5500 (20°C, 589 nm)
- GC:≥97.5 %