Order Entry
Export
ContactUsLinkComponent
Piperine 98%
Piperine 98%
Certificates
undefined
Piperine 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Off-white to yellow to tan Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    128 to 132 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information