Calcium carbonate, heavy powder USP
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Marble
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
Density:2.93 g/cm³ (20 °C)
Molecular Weight:100.09 g/mol
Melting Pt:825
Merck Index:13,01658
Storage Temperature:Ambient
EINECS:207-439-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98% Min.