Order Entry
Export
ContactUsLinkComponent
 
Anti-SPR Antibody Pair
  ABNOH00006697-PW1
 :  Abnova
Anti-SPR Antibody Pair
  ABNOH00006697-PW1
 :  Abnova

 

  • Antigen name:
    Surface Plasmon Resonance
  • Antigen symbol:
    SPR
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    6697
  • Antigen synonyms:
    SPRE_HUMAN.|SDR38C1|Sepiapterin reductase 7,8 dihydrobiopterin:NADP+ oxidoreductase|member 1|Short chain dehydrogenase/reductase family 38C|SPR
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-SPR (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-SPR (50 µl)

Type:
Antigen: SPR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human