Anti-RFC1 Rabbit Polyclonal Antibody
Catalog # ABGEAP19110A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Replication factor C subunit 1
- Antigen symbol:RFC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Gene ID:5981
- Antigen synonyms:RECC1|A1|MHCBFB|RFC|RFC140|PO-GA
- Amino acid number:26 to 54
- Storage buffer:pH 7,4
- Molecular weight:128.255 kDa
- Sequence:KEIKVNSSRKEDDFC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.3
- Shipping temperature:4 °C
- Immunogen:This RFC1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human RFC1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RFC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human