Specifications
- Antibody type:Primary
- Antigen name:Autophagy protein 5
- Antigen symbol:APG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:APG5|APG5-like|APG5L|Autophagy protein 5|APG 5|Apoptosis-specific protein|ASP|ATG5
- Amino acid number:82 to 97
- UniProtKB:Q9H1Y0
- Sequence:DRFDQFWAINRKLMEY
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human APG5L (82-97aa DRFDQFWAINRKLMEY), identical to the related mouse sequence, and different from the related rat sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Autophagy protein 5(ATG5) detection. Tested with WB in Human;Mouse;Rat.
Autophagy protein 5 is a protein that in humans is encoded by the ATG5 gene. It is also known as APG5 or ASP, and this gene is mapped to 6q21. It is found that knockdown of ATG5 in hepatocytes increased triglyceride levels with oleate or a second endogenous stimulus for triglyceride formation. These hepatocytes with ATG5 knockdown also had increased lipid droplet number and size. ATG5 is an E3 ubiquitin ligase which is necessary for autophagy due to its role in autophagosome elongation. It is activated by ATG7 and forms a complex with ATG12 and ATG16L1. This complex is necessary for LC3-1 conjugation to PE to form LC3-II.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: APG5L/ATG5
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat