Anti-Melatonin Rat Polyclonal Antibody
USBIM2871-04F
- Antibody type:Primary
- Antigen name:Melatonin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by protein carrier (BSA) affinity column, and ammonium sulphate precipitation
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Melatonin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: