Anti-Heat Shock Protein 90, beta Mouse Monoclonal Antibody [clone: 10F733]
Catalog # USBIH1832-98D1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 90 beta
- Antigen symbol:HSP90B
- Clonality:Monoclonal
- Clone:10F733
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot:1 ?g/ml
Type: Primary
Antigen: Heat Shock Protein 90, beta
Clonality: monoclonal
Clone: 10F733
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse