Anti-CD137 Ligand Mouse Monoclonal Antibody (Biotin) [clone: 7H151]
Catalog # USBIC2548-55B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD137 Ligand
- Antigen symbol:CD137L
- Clonality:Monoclonal
- Clone:7H151
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,5, 0,2% BSA, 0,04% sodium azide,5% glycerol
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD137 Ligand
Clonality: Monoclonal
Clone: 7H151
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human