Anti-ADAM10 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 9H29]
USBIA0859-20E
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 10
- Antigen symbol:ADAM10
- Clonality:Monoclonal
- Clone:9H29
- Conjugation:FAM (Carboxyfluorescein)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:214 to 672
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: ADAM10 (ADAM metallopeptidase domain 10)
Clonality: monoclonal
Clone: 9H29
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human