Anti-F12 Rabbit Polyclonal Antibody
The Factor XII Antibody from Novus Biologicals is a rabbit polyclonal antibody to Factor XII. This antibody reacts with human, mouse, rat, primate. The Factor XII Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: Factor XII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate
- Catalog No:
- NOVUNBP1-94203
- NOVUNBP1-94203SS
- Antigen symbol:
- F12
- F12
- Antigen name:
- Coagulation Factor 12
- Coagulation Factor 12
- Antigen synonyms:
- HAF|Coagulation Factor XII|HAE3|Hageman Factor|HAEX
- HAF|Coagulation Factor XII|HAE3|Hageman Factor|HAEX
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2161
- 2161
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice