Anti-LOX Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB100-2530SS
- NOVUNB100-2530
- Antigen symbol:
- LOX
- LOX
- Antigen name:
- Lysyl oxidase
- Lysyl oxidase
- Antigen synonyms:
- Lysyl oxidase|Protein-lysine-6 oxidase|LOX
- Lysyl oxidase|Protein-lysine-6 oxidase|LOX
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4015
- 4015
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice