Anti-MK2 Goat Polyclonal Antibody
Catalog # BIRBORB107653-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:MK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region (near C Terminus)
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:128000
Type: Primary
Antigen: MK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: