Order Entry
Export
ContactUsLinkComponent
Anti-TSSK3 Mouse Monoclonal Antibody [clone: 3G8]
Anti-TSSK3 Mouse Monoclonal Antibody [clone: 3G8]
Catalog # ABNOH00081629-M02
Supplier:  Abnova
undefined
Anti-TSSK3 Mouse Monoclonal Antibody [clone: 3G8]
Catalog # ABNOH00081629-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Testis-specific serine kinase 3
  • Antigen symbol:
    TSSK3
  • Clonality:
    Monoclonal
  • Clone:
    3G8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    81629
  • Antigen synonyms:
    SPOGA3|STK22D|TSK3|STK22C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: TSSK3
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human