Anti-GEMIN4 Mouse Monoclonal Antibody [clone: 3E1]
ABNOH00050628-M01
: Abnova
- Antibody type:Primary
- Antigen name:Gemin 4
- Antigen symbol:GEMIN4
- Clonality:Monoclonal
- Clone:3E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:50628
- Antigen synonyms:Component of gems 4|HCAP1|Gem associated protein 4|GEMI4_HUMAN.|HC56|Gemin-4|HHRF 1|Gemin4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GEMIN4
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human