Anti-IL36B Mouse Monoclonal Antibody [clone: 1E4]
Catalog # ABNOH00027177-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 36 Beta
- Antigen symbol:IL36B
- Clonality:Monoclonal
- Clone:1E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:27177
- Antigen synonyms:IL-1H2|FIL1|FIL1H|IL1F8|IL-1F8|IL1H2|IL1-ETA|FIL1-(ETA)|FILI-(ETA)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IL36B
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human