Order Entry
Export
ContactUsLinkComponent
Anti-GOLGA5 Mouse Monoclonal Antibody [clone: 6B3]
Anti-GOLGA5 Mouse Monoclonal Antibody [clone: 6B3]
Catalog # ABNOH00009950-M01
Supplier:  Abnova
undefined
Anti-GOLGA5 Mouse Monoclonal Antibody [clone: 6B3]
Catalog # ABNOH00009950-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Golgin Subfamily A Member 5
  • Antigen symbol:
    GOLGA5
  • Clonality:
    Monoclonal
  • Clone:
    6B3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    9950
  • Antigen synonyms:
    GOLGA5|ret-II|PIG31|Golgin subfamily A member 5|GOLIM5|Cell proliferation-inducing gene 31 protein|RETII|RFG5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: GOLGA5
Clonality: Monoclonal
Clone: 6B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human