Anti-EIF3J Mouse Polyclonal Antibody
ABNOH00008669-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 3, subunit J
- Antigen symbol:EIF3J
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8669
- Antigen synonyms:eIF3-alpha|EIF3S1|eIF3-p35
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: EIF3J
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human