Anti-UROD Mouse Monoclonal Antibody [clone: 2F5]
ABNOH00007389-M08
: Abnova
- Antibody type:Primary
- Antigen name:Uroporphyrinogen decarboxylase
- Antigen symbol:UROD
- Clonality:Monoclonal
- Clone:2F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7389
- Antigen synonyms:URO D|Uroporphyrinogen decarboxylase|Uroporphyrinogen III decarboxylase.|UPD|PCT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: UROD
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human