Anti-FRZB Mouse Monoclonal Antibody [clone: 3F4]
Catalog # ABNOH00002487-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Frizzled-Related Protein
- Antigen symbol:FRZB
- Clonality:Monoclonal
- Clone:3F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2487
- Antigen synonyms:FRZB-PEN|hFIZ|SFRP3|FRP-3|OS1|FRE|FRITZ|FZRB|FRZB1|SRFP3|FRZB-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FRZB
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human