Hand disinfectants, Sterillium® Med
Supplier: Bode
Certificates
About this item
Sterillium® med is an ethanol based hand disinfectant designed for hygienic and surgical rub-in hand disinfection with good skin tolerability.
- Active against bacteria, fungi and enveloped viruses
- 1,5 min of application time for surgical hand disinfection
- Independent of washbasin and water
- Colourant- and fragrance-free
Caution: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.
* These articles are only available in Germany.