ESD Safety shoes, lace-up, Alucap, Bayreuth, 2205
Supplier: SCHURR SCHUHVERTRIEB
Certificates
About this item
ESD safety shoe made from microfibre Cordura®, DRYtech® lining and a PU sole profile 'Max'.
- Antibacterial and breathable DRYtech® textile lining
- Removable insole made from PUR-foam
- Orthopaedic insoles possible
- Triple shock absorber at heel and ball of the foot relieves joints
- Protective aluminium toe cap (200 J)
For ESD and kitchen use.
Colour: White
Width: 11
Weight: 510 g
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRA