ContactUsLinkComponent
Avantor Fluid HandlingChemical & Material Compatibility DatabaseMasterflex Pump Head Roller Material Compatibility DatabaseMasterflex Tubing and Food Compatibility DatabaseMasterflex Tubing Compatibility Database
Chemical & Material Compatibility Database
Use the dropdowns below to search by Chemical , Material or both.
Shop Avantor’s chemical portfolio
Including our high purity J.T.Baker brand and versatile Macron Fine Chemicals brand.
Contact Us
Our team of expertsre here to help
WARNING
The information in this chart has been supplied to Masterflex by other reputable sources and is to be used ONLY as a guide in selecting equipment for appropriate chemical compatibility. Before permanent installation, test the equipment with the chemicals and under the specific conditions of your application.
Ratings of chemical behavior listed in this chart apply at a 48-hr exposure period. Masterflex has no knowledge of possible effects beyond this period. Masterflex does not warrant (neither express nor implied) that the information in this chart is accurate or complete or that any material is suitable for any purpose.
DANGER
Variations in chemical behavior during handling due to factors such as temperature, pressure, and concentrations can cause equipment to fail, even though it passed an initial test.
SERIOUS INJURY MAY RESULT
Use suitable guards and/or personal protections when handling chemicals.