Masterflex® L/S® Replacement Fuse, Slo-Blo, 1.6 A/250 V, 5 X 20 mm
Catalog # MFLX77500-11
Supplier: Avantor Fluid Handling
About this item
Make the most of your drive with these replacement parts.
- Optimize performance of your Masterflex pumps
Suitable for use with Masterflex® B/T® drive 77111-30.
For use with the following Masterflex L/S drives: 07520-47, 07520-57, and 07520-67; 07523-47 and 07523-57; 07524-45 and 07524-55; 07553-75, 07553-76, 07553-77, 07553-85, and 07553-57; 77300-25 and 77300-35; 77521-47 and 77521-57.
- Description:Replacement Fuse, 1,6 A/250 V, 5 X 20 mm, Slo-Blo
- Environmentally Preferable:
- Compatible with:Masterflex L/S Drives
- Pk:2
Frequently Bought Together