2,4-Hexadiene mixture of isomers ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Hexa-2,4-diene
CAS Number:592-46-1
:
Formula:C₆H₁₀
MDL Number:MFCD00009297
Boiling Pt:83°C
Molecular Weight:82.15 g/mol
Melting Pt:-79
Flash Pt:–8°C
EINECS:209-757-3
UN:2458
ADR:3 II
:GHS02
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomer)
Specification Test Results
Frequently Bought Together